Invitrogen
Manufacturer Code:PA5106983
Catalog # PIPA5106983
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide located within the following region: AYYFSRDDVA LNNFSRYFLH QSREETEHAE KLMRLQNQRG GRIRLQDIKK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ferritin H subunit; Ferritin, mitochondrial; mitochondrial ferritin
Gene Aliases: FTMT; MTF
UniProt ID: (Human) Q8N4E7
Entrez Gene ID: (Human) 94033
Molecular Function:
storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.