Invitrogen
Manufacturer Code:PA548073
Catalog # PIPA548073
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHO-derived recombit mouse Follistatin-like 4/FSTL4 Trp23-Val841 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: follistatin-like 4; Follistatin-like protein 4; Follistatin-related protein 4; FSL4 FSTL4 follistatin-like 4 Follistatin-like protein 4 follistatin-related protein 4 KIAA1061; m-D/Bsp120I 1-1
Gene Aliases: B230374F23Rik; FSTL4; KIAA1061; mKIAA1061; SPIG1
UniProt ID: (Human) Q6MZW2, (Mouse) Q5STE3
Entrez Gene ID: (Human) 23105, (Rat) 303130, (Mouse) 320027
Molecular Function:
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.