Invitrogen
Manufacturer Code:PA579285
Catalog # PIPA579285
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of human FSTL3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: follistatin-like 3 (secreted glycoprotein); Follistatin-like protein 3; Follistatin-related gene protein; Follistatin-related protein 3; Follistatin-related protein 3 Follistatin-like protein 3 Follistatin-related gene protein FSTL3 FLRG UNQ674/PRO1308
Gene Aliases: E030038F23Rik; FLRG; FSRP; FSTL3; UNQ674/PRO1308
UniProt ID: (Human) O95633, (Mouse) Q9EQC7, (Rat) Q99PW7
Entrez Gene ID: (Human) 10272, (Mouse) 83554, (Rat) 114031
Molecular Function: enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.