Proteintech
Manufacturer Code:201821AP150UL
Catalog # 50-173-2753
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FSTL1 Fusion Protein Ag14057 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: follistatin like 1 Follistatin like protein 1 Follistatin related protein 1 FRP FSL1 FSTL1 TSC-36; follistatin-like 1; Follistatin-like protein 1; Follistatin-related protein 1
Gene Aliases: FRP; FSL1; Fstl; FSTL1; MIR198; OCC-1; OCC1; tsc36
UniProt ID: (Human) Q12841, (Rat) Q62632
Entrez Gene ID: (Human) 11167, (Rat) 79210
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.