Novus Biologicals
Manufacturer Code:NBP254329UV
Catalog # NBP254329UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | FSHR/1400 |
Immunogen | Recombinant human full-length FSHR protein |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: follicle stimulating hormone receptor follicle-stimulating hormone receptor Follitropin receptor FSH receptor FSH-R LGR1FSHRO MGC141667 MGC141668 ODG1; Follicle-stimulating hormone receptor; Follitropin receptor; FSH receptor; FSH-R
Gene Aliases: FSHR; FSHR1; FSHRO; LGR1; ODG1
UniProt ID: (Human) P23945
Entrez Gene ID: (Human) 2492
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.