Abnova
Manufacturer Code:MAB14440
Catalog # 89-034-523
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | FSCN1/417 |
Immunogen | Recombinant protein corresponding to full length human FSCN1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 55 kDa actin-bundling protein; Fascin; fascin homolog 1, actin-bundling protein; FLJ38511; p55; singed-like (fascin homolog, sea urchin); Singed-like protein; SNL
Gene Aliases: FAN1; FSCN1; HSN; p55; SNL
UniProt ID: (Human) Q16658
Entrez Gene ID: (Human) 6624
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.