Invitrogen
Manufacturer Code:PA547793
Catalog # PIPA547793
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse sFRP-3 Ala33-Ser323 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Frezzled; Fritz; Fritz FRZB Frzb1 FIZ FREOS1 Frezzled Fritz FRITZ frizzled-related protein Frizzled-related protein 1 FRP FRP-3 FrzB-1 FRZB-1 FRZB1sFRP-3 FRZB-PEN FZRB hFIZ secreted frizzled-related protein 3 SFRP3frezzled SRFP3frizzled homolog-related; Frizzled-related protein 1; FrzB-1; Secreted frizzled-related protein 3; secreted frizzled-related sequence protein 3; sFRP-3
Gene Aliases: Fiz; Fre; frezzled; fritz; Frp; Frzb; frzb-1; Frzb1; Sfrp3
UniProt ID: (Mouse) P97401
Entrez Gene ID: (Mouse) 20378
Molecular Function:
G-protein coupled receptor
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.