Invitrogen
Manufacturer Code:PA544188
Catalog # PIPA544188
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human FRZB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Frezzled; Fritz; frizzled homolog-related; Frizzled-related protein 1; Frzb; FrzB-1; Secreted frizzled-related protein 3; secreted frizzled-related sequence protein 3; sFRP-3
Gene Aliases: cb291; FIZ; FRE; frezzled; FRITZ; FRP; FRP-3; FRZB; FRZB-1; FRZB-PEN; FRZB1; fzb-1; fzb1; FZRB; hFIZ; id:ibd5075; ik:tdsubc_2e7; OS1; SFRP3; SRFP3; xx:tdsubc_2e7
UniProt ID: (Human) Q92765, (Bovine) Q95117, (Mouse) P97401
Entrez Gene ID: (Rabbit) 100008629, (Human) 2487, (Bovine) 281170, (Horse) 100068206, (Dog) 478827, (Mouse) 20378, (Guinea pig) 100716120, (Rat) 295691, (Zebrafish) 30119
Molecular Function:
G-protein coupled receptor
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.