Invitrogen
Manufacturer Code:PA5117059
Catalog # PIPA5117059
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 166-325 of human FRZB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fiz Fre FREOS1 frezzled Fritz frizzled homolog-related frizzled-related protein frizzled-related protein 1 FRP FRP-3 FRZB FRZB1 FrzB-1 FRZB1sFRP-3 FRZB-PEN FZRB hFIZ OS1 Secreted frizzled-related protein 3 secreted frizzled-related sequence protein 3 Sfrp3 sFRP-3 SFRP3frezzled SRFP3; Frezzled; Fritz; frizzled homolog-related; Frizzled-related protein 1; FrzB-1; Secreted frizzled-related protein 3; secreted frizzled-related sequence protein 3; sFRP-3
Gene Aliases: FIZ; FRE; frezzled; FRITZ; FRP; FRP-3; FRZB; FRZB-1; FRZB-PEN; FRZB1; FZRB; hFIZ; OS1; SFRP3; SRFP3
UniProt ID: (Human) Q92765, (Mouse) P97401
Entrez Gene ID: (Human) 2487, (Mouse) 20378
Molecular Function: G-protein coupled receptor receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.