Novus Biologicals
Manufacturer Code:NBP2298860.05MG
Catalog # NB019500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | T5 |
Immunogen | Peptide corresponding to residues 283-297 of human FRS2/SNT-1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FGFR signalling adaptor; FGFR signalling adaptor FGFR-signaling adaptor SNT fibroblast growth factor receptor substrate 2 FRS2A FRS2alpha SNT SNT1 SNT-1FGFR substrate 2 Suc1-associated neurotrophic factor target 1; FGFR substrate 2; FGFR-signaling adaptor SNT; Fibroblast growth factor receptor substrate 2; SNT-1; Suc1-associated neurotrophic factor target 1
Gene Aliases: FRS1A; FRS2; FRS2A; FRS2alpha; SNT; SNT-1; SNT1
UniProt ID: (Human) Q8WU20
Entrez Gene ID: (Human) 10818
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.