Abnova
Manufacturer Code:PAB20970
Catalog # 89-122-467
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human FRRS1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant FRRS1.
For Research Use Only
Protein Aliases: Ferric-chelate reductase 1; SDFR2; SDR-2; SDR2; stromal cell derived factor receptor 2 homolog; Stromal cell-derived receptor 2
Gene Aliases: FRRS1; SDFR2; SDR2
UniProt ID: (Human) Q6ZNA5
Entrez Gene ID: (Human) 391059
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.