Novus Biologicals
Manufacturer Code:NBP259102
Catalog # NBP259102
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Maps to a region between 572 and 622 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4.1 ezrin radixin moesin (FERM)-containing protein; expanded homolog; FERM domain containing 6; FERM domain-containing protein 6; Willin
Gene Aliases: c14_5320; C14orf31; EX1; FRMD6; Willin
UniProt ID: (Human) Q96NE9
Entrez Gene ID: (Human) 122786
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.