OriGene
Manufacturer Code:TA500526
Catalog # 50-167-1662
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6D2 |
Immunogen | Full length human recombit protein of human FRK produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FYN-related kinase; fyn-related Src family tyrosine kinase; GTK PTK5 RAK; Nuclear tyrosine protein kinase RAK; Protein-tyrosine kinase 5; PTK5 protein tyrosine kinase 5; Tyrosine-protein kinase FRK
Gene Aliases: FRK; GTK; PTK5; RAK
UniProt ID: (Human) P42685
Entrez Gene ID: (Human) 2444
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.