Novus Biologicals
Manufacturer Code:NBP271351IR
Catalog # NBP271351IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI12F9 |
Immunogen | Full-length protein expressed in 293T cell transfected with human FRA-1/FOSL1 expression vector. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: FOS like 1, AP-1 trancription factor subunit; FOS like antigen 1; FOS-like antigen 1; FOS-like antigen 1 FOS-like antigen-1 fos-related antigen 1 FRA FRA1 FRA-1; FOS-like antigen-1; Fos-related antigen 1; FRA-1
Gene Aliases: FOSL1; FRA; fra-1; FRA1
UniProt ID: (Human) P15407
Entrez Gene ID: (Human) 8061
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.