Invitrogen
Manufacturer Code:700914
Catalog # PI700914
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 5H10L18 |
Immunogen | proprietary |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Forkhead box protein P3; Forkhead box protein P3 41 kDa form; Forkhead box protein P3, C-terminally processed; FOXP3delta7; immune dysregulation, polyendocrinopathy, enteropathy, X-linked; immunodeficiency, polyendocrinopathy, enteropathy, X-linked; JM2 AIID DIETER IPEX MGC141961 MGC141963 PIDX XPID FOXP3delta7 forkhead box protein P3 immune dysregulation polyendocrinopathy enteropathy X-linked immunodeficiency polyendocrinopathy enteropathy X-linked scurfin; Scurfin; scurfy
Gene Aliases: AIID; DIETER; FOXP3; IPEX; JM2; PIDX; scurfin; sf; XPID
UniProt ID: (Human) Q9BZS1, (Mouse) Q99JB6
Entrez Gene ID: (Human) 50943, (Mouse) 20371
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.