Invitrogen
Manufacturer Code:MA531419
Catalog # PIMA531419
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL5312 |
Immunogen | Recombit protein corresponding to Human FOXP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CAG repeat protein 44; CAGH44 SPCH1 TNRC10; Forkhead box protein P2; forkhead/winged-helix transcription factor; trinucleotide repeat containing 10; Trinucleotide repeat-containing gene 10 protein
Gene Aliases: 2810043D05Rik; AI449000; CAG-16; CAGH44; D0Kist7; FOXP2; RGD1559697; SPCH1; TNRC10
UniProt ID: (Human) O15409, (Mouse) P58463, (Rat) P0CF24
Entrez Gene ID: (Human) 93986, (Mouse) 114142, (Rat) 500037
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.