Novus Biologicals
Manufacturer Code:NBP245712
Catalog # NBP245712
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A4 |
Immunogen | Human recombinant protein fragment corresponding to amino acids 206-505 of human FOXO4 (NP_005929) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AFX AFX1myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog)translocated to 7 Fork head domain transcription factor AFX1 forkhead box O4 forkhead box protein O4 MLLT7MGC120490 myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog Drosophila)translocated to 7; Fork head domain transcription factor AFX1; Forkhead box protein O4; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 7
Gene Aliases: AFX; AFX1; FOXO4; MLLT7
UniProt ID: (Human) P98177
Entrez Gene ID: (Human) 4303
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.