Invitrogen
Manufacturer Code:PA585078
Catalog # PIPA585078
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human FOXL2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BPES antibody BPES1 antibody PFRK antibody PINTO antibody POF3 antibody FOXL2 antibody forkhead transcription factor FOXL2 antibody forkhead box protein L2 antibody forkhead box L2 antibody; Forkhead box protein L2; P-Frk; Pituitary forkhead factor; putative transcription factor foxl2
Gene Aliases: AU045128; BPES; Foxl2; Pfrk; PINTO
UniProt ID: (Mouse) O88470
Entrez Gene ID: (Mouse) 26927
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.