Invitrogen
Manufacturer Code:PA131950
Catalog # PIPA131950
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide DSKTGALHSRLDL corresponding to amino acids 364-376 of FOX L2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: forkhead box L2; forkhead box L2 forkhead transcription factor FOXL2; Forkhead box protein L2; forkhead transcription factor FOXL2; forkhead transcription factor L2; P-Frk; Pituitary forkhead factor; putative transcription factor foxl2
Gene Aliases: AU045128; BPES; BPES1; FOXL2; PFRK; PINTO; POF3
UniProt ID: (Bovine) Q6VFT7, (Human) P58012, (Mouse) O88470
Entrez Gene ID: (Bovine) 281770, (Human) 668, (Mouse) 26927, (Rat) 367152
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.