Novus Biologicals
Manufacturer Code:NBP222473
Catalog # NBP222473
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic Peptide corresponding to residues M(1) M A S Y P E P E D T A G T(14) of mouse FoxL2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BPES BPES1PINTO forkhead box L2 forkhead box protein L2 forkhead transcription factor FOXL2 PFRK POF3; Forkhead box protein L2; forkhead transcription factor FOXL2
Gene Aliases: BPES; BPES1; FOXL2; PFRK; PINTO; POF3
UniProt ID: (Human) P58012
Entrez Gene ID: (Human) 668
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.