Novus Biologicals
Manufacturer Code:NBP270013PECY7
Catalog # N27013PECY7
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to a C-terminal portion of the human FOXL2 protein (between amino acids 350-376) [UniProt P58012] |
Conjugate | PE-Cyanine7 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Rabbit Polyclonal Antibody
Protein Aliases: BPES BPES1PINTO forkhead box L2 forkhead box protein L2 forkhead transcription factor FOXL2 PFRK POF3; Forkhead box protein L2; forkhead transcription factor FOXL2
Gene Aliases: BPES; BPES1; FOXL2; PFRK; PINTO; POF3
UniProt ID: (Human) P58012
Entrez Gene ID: (Human) 668
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.