Abnova
Manufacturer Code:H00002300M10
Catalog # 89-022-139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C3 |
Immunogen | FOXL1 (NP_005241 132 a.a. ~ 240 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant FOXL1.
For Research Use Only
Protein Aliases: FKH6; FKHL11; Forkhead box protein L1; forkhead-like 11; Forkhead-related protein FKHL11; Forkhead-related transcription factor 7; FREAC-7; FREAC7
Gene Aliases: FKH6; FKHL11; FOXL1; FREAC7
UniProt ID: (Human) Q12952
Entrez Gene ID: (Human) 2300
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.