Invitrogen
Manufacturer Code:PA599303
Catalog # PIPA599303
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human FOXL1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FKH6 fkh-6 Fkhl11 forkhead box L1 forkhead box protein L1 forkhead homolog 6 forkhead-like 11 forkhead-related protein FKHL11 Forkhead-related transcription factor 7 FOXL1 FREAC7 FREAC-7 Transcription factor FKH-6; Forkhead box protein L1; forkhead-like 11; Forkhead-related protein FKHL11; Forkhead-related transcription factor 7; FREAC-7
Gene Aliases: FKH6; FKHL11; FOXL1; FREAC7
UniProt ID: (Human) Q12952
Entrez Gene ID: (Human) 2300
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.