Invitrogen
Manufacturer Code:PA526220
Catalog # PIPA526220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 94-119 amino acids from the central region of human FOXL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FKH6 FKHL11 FREAC7 FREAC-7 forkhead box protein L1 forkhead-like 11 forkhead-related protein FKHL11 forkhead-related transcription factor 7; Forkhead box protein L1; forkhead homolog 6; forkhead-like 11; Forkhead-related protein FKHL11; Forkhead-related transcription factor 7; FREAC-7; Transcription factor FKH-6
Gene Aliases: fkh-6; FKH6; FKHL11; FOXL1; FREAC7
UniProt ID: (Human) Q12952, (Mouse) Q64731
Entrez Gene ID: (Human) 2300, (Mouse) 14241
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.