Invitrogen
Manufacturer Code:14996595
Catalog # 14-996-595
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2A5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FKHL13 FKHL-13 fork head homologue 4 forkhead box J1 forkhead box protein J1 forkhead transcription factor HFH-4 forkhead-like 13 forkhead-related protein FKHL13 Foxj1 Hepatocyte nuclear factor 3 forkhead homolog 4 HFH4 HFH-4 HNF-3/forkhead homolog 4 MGC35202; fork head homologue 4; Forkhead box protein J1; forkhead transcription factor HFH-4; forkhead-like 13; Forkhead-related protein FKHL13; Hepatocyte nuclear factor 3 forkhead homolog 4; HFH-4; HNF-3/forkhead homolog 4
Gene Aliases: FKHL-13; FKHL13; FOXJ1; HFH-4; HFH4
UniProt ID: (Human) Q92949, (Mouse) Q61660
Entrez Gene ID: (Human) 2302, (Mouse) 15223
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.