Invitrogen
Manufacturer Code:PA568947
Catalog # PIPA568947
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse FOXJ1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: fork head homologue 4; Forkhead box protein J1; Forkhead box protein J1 Forkhead-related protein FKHL13 Hepatocyte nuclear factor 3 forkhead homolog 4 HFH-4 FOXJ1 FKHL13 HFH4; forkhead transcription factor HFH-4; forkhead-like 13; Forkhead-related protein FKHL13; Hepatocyte nuclear factor 3 forkhead homolog 4; HFH-4; HNF-3/forkhead homolog 4
Gene Aliases: FKHL-13; FKHL13; FOXJ1; HFH-4; HFH4
UniProt ID: (Human) Q92949, (Mouse) Q61660, (Rat) Q63247
Entrez Gene ID: (Dog) 475917, (Human) 2302, (Horse) 100147129, (Bovine) 505891, (Rabbit) 100008873, (Guinea pig) 100722854, (Mouse) 15223, (Rat) 116557
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.