Invitrogen
Manufacturer Code:PA536210
Catalog # PIPA536210
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 264-315 of human FoxJ1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: fork head homologue 4; Forkhead box protein J1; Forkhead box protein J1 Forkhead-related protein FKHL13 Hepatocyte nuclear factor 3 forkhead homolog 4 HFH-4 FOXJ1 FKHL13 HFH4; forkhead transcription factor HFH-4; forkhead-like 13; Forkhead-related protein FKHL13; Hepatocyte nuclear factor 3 forkhead homolog 4; HFH-4; HNF-3/forkhead homolog 4
Gene Aliases: FKHL-13; FKHL13; FOXJ1; HFH-4; HFH4
UniProt ID: (Human) Q92949, (Mouse) Q61660, (Rat) Q63247
Entrez Gene ID: (Human) 2302, (Mouse) 15223, (Rat) 116557
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.