Invitrogen
Manufacturer Code:PA541472
Catalog # PIPA541472
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human FOXH1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fast-1; Fast-2; Forkhead activin signal transducer 1; Forkhead activin signal transducer 2; forkhead activin signal transducer-1; Forkhead box H1 FAST-1 FAST1; Forkhead box protein H1; hFAST-1; TGF-beta/activin signal transducer
Gene Aliases: FAST-1; FAST1; FAST2; FOXH1
UniProt ID: (Human) O75593
Entrez Gene ID: (Pig) 100154027, (Human) 8928, (Horse) 100064793, (Bovine) 493733, (Rat) 300054, (Guinea pig) 100716330
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.