Invitrogen
Manufacturer Code:PA541493
Catalog # PIPA541493
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human FOXG1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BF-1; BF-2; BF1; Brain factor 1; Brain factor 2; Forkhead box G1 BF1 BF2 QIN FKH2 HBF2 HFK1 HFK2 HFK3 KHL2 FHKL3 FKHL1 FKHL2 FKHL3 FKHL4 HBF-1 HBF-2 HBF-3 FOXG1A FOXG1B FOXG1C HBF-G2; Forkhead box protein G1; Forkhead box protein G1A; Forkhead box protein G1B; Forkhead box protein G1C; forkhead-like 1; forkhead-like 2; forkhead-like 3; forkhead-like 4; forkhead-like transcription factor BF-1; Forkhead-related protein FKHL1; Forkhead-related protein FKHL2; Forkhead-related protein FKHL3; FoxG1; HFK1; HFK2; HFK3; HNF-3/forkhead homolog, brain factor 1; oncogene QIN; transcription factor BF-1
Gene Aliases: 2900064B05Rik; BF-1; BF1; BF1A; BF2; FHKL3; FKH2; FKHL1; FKHL2; FKHL3; FKHL4; FOXG1; FOXG1A; FOXG1B; FOXG1C; HBF-1; HBF-2; HBF-3; HBF-G2; HBF2; Hfh9; Hfhbf1; HFK1; HFK2; HFK3; KHL2; QIN; RATBF1A
UniProt ID: (Human) P55316, (Rat) Q00939, (Mouse) Q60987
Entrez Gene ID: (Human) 2290, (Rat) 24370, (Mouse) 15228
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.