Novus Biologicals
Manufacturer Code:NBP281942
Catalog # NB017253
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FOXG1 antibody was raised against an 18 amino acid peptide near the center of human FOXG1. The immunogen is located within amino acids 160 - 210 of FOXG1. Amino Acid Squence: KEGEKKNGKYEKPPFSYN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BF-1; BF-1 BF1HBF-2 BF2 BF-2 Brain factor 1 Brain factor 2 FHKL3 FKHL1forkhead-like 1 FKHL2 FKHL3 FKHL4 forkhead box G1 forkhead box G1A forkhead box G1B forkhead box G1C Forkhead box protein G1A Forkhead box protein G1B Forkhead box protein G1C forkhead-like 3 forkhead-like 4 Forkhead-related protein FKHL1 Forkhead-related protein FKHL2 Forkhead-related protein FKHL3 FOXG1A FOXG1Bforkhead-like 2 FOXG1C HBF-1 hBF-2 HBF-3 HFK1HBF-G2 HFK2HBF2 HFK3KHL2 oncogene QIN QINFKH2forkhead box protein G1; BF-2; Brain factor 1; Brain factor 2; Forkhead box protein G1; Forkhead box protein G1A; Forkhead box protein G1B; Forkhead box protein G1C; forkhead-like 1; forkhead-like 2; forkhead-like 3; forkhead-like 4; Forkhead-related protein FKHL1; Forkhead-related protein FKHL2; Forkhead-related protein FKHL3; HFK1; HFK2; HFK3; oncogene QIN
Gene Aliases: BF1; BF2; FHKL3; FKH2; FKHL1; FKHL2; FKHL3; FKHL4; FOXG1; FOXG1A; FOXG1B; FOXG1C; HBF-1; HBF-2; HBF-3; HBF-G2; HBF2; HFK1; HFK2; HFK3; KHL2; QIN
UniProt ID: (Human) P55316
Entrez Gene ID: (Human) 2290
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.