Invitrogen
Manufacturer Code:PA5113636
Catalog # PIPA5113636
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of mouse Foxf1a |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACDMPV AI450827 Fkhl5 forkhead box F1 forkhead box F1a forkhead box F1a-like Forkhead box protein F1 Forkhead drosophila homolog-like 5 Forkhead-related activator 1 Forkhead-related protein FKHL5 Forkhead-related transcription factor 1 FOXF1 Foxf1a Freac1 FREAC-1 Hepatocyte nuclear factor 3 forkhead homolog 8 Hfh8 HFH-8 HNF-3/forkhead homolog 8 LOC100341838; forkhead box F1a; Forkhead box protein F1; Forkhead-related protein FKHL5; Forkhead-related transcription factor 1; FREAC-1; Hepatocyte nuclear factor 3 forkhead homolog 8; HFH-8; HNF-3/forkhead homolog 8
Gene Aliases: AI450827; Fkhl5; Foxf1; Foxf1a; Freac-1; FREAC1; HFH-8; Hfh8
UniProt ID: (Mouse) Q61080
Entrez Gene ID: (Mouse) 15227
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.