Invitrogen
Manufacturer Code:PA575289
Catalog # PIPA575289
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 64-116 of Human FoxE3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Forkhead box protein E3; Forkhead box protein E3 Forkhead-related protein FKHL12 Forkhead-related transcription factor 8 FREAC-8 FKHL12 FREAC8 FOXE3; forkhead, drosophila, homolog-like 12; Forkhead-related protein FKHL12; Forkhead-related transcription factor 8; FREAC-8; Hepatocyte nuclear factor 3 forkhead homolog 7; HFH-7
Gene Aliases: FKHL12; FOXE3; FREAC-8; FREAC8; HFH7; HNF3
UniProt ID: (Human) Q13461, (Rat) Q63250
Entrez Gene ID: (Human) 2301, (Rat) 171302
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.