Invitrogen
Manufacturer Code:PA5101159
Catalog # PIPA5101159
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human FOXE3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dyl dysgenetic lens FKHL12 forkhead box E3 forkhead box protein E3 forkhead drosophila homolog-like 12 forkhead-related protein FKHL12 Forkhead-related transcription factor 8 FOXE3 FREAC8 FREAC-8 Hepatocyte nuclear factor 3 forkhead homolog 7 Hfh7 HFH-7 HNF3 rct Rinshoken cataract; dysgenetic lens; Forkhead box protein E3; forkhead, drosophila, homolog-like 12; Forkhead-related protein FKHL12; Forkhead-related transcription factor 8; FREAC-8; Hepatocyte nuclear factor 3 forkhead homolog 7; HFH-7; Rinshoken cataract
Gene Aliases: dyl; FKHL12; FOXE3; FREAC-8; FREAC8; HFH7; HNF3; rct
UniProt ID: (Human) Q13461, (Rat) Q63250, (Mouse) Q9QY14
Entrez Gene ID: (Human) 2301, (Rat) 171302, (Mouse) 30923
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.