Novus Biologicals
Manufacturer Code:NB1001273
Catalog # NB1001273
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide: AGRSDMDPPAAFS representing the N-Terminus of the human protein (residues 2-14) according to NP_036318. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ASMD FKHL12forkhead drosophila homolog-like 12 forkhead box E3 Forkhead-related protein FKHL12 Forkhead-related transcription factor 8 FREAC-8 FREAC8forkhead box protein E3; Forkhead box protein E3; forkhead, drosophila, homolog-like 12; Forkhead-related protein FKHL12; Forkhead-related transcription factor 8; FREAC-8
Gene Aliases: FKHL12; FOXE3; FREAC8
UniProt ID: (Human) Q13461
Entrez Gene ID: (Human) 2301
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.