Invitrogen
Manufacturer Code:PA527143
Catalog # PIPA527143
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 311 and 373 of FOXE1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FKHL15 FOXE2 HFKH4 HFKL5 TITF2 TTF-2 TTF2 HNF-3/fork head-like protein 5 forkhead box E2 forkhead box protein E1 forkhead box protein E2 forkhead drosophila homolog-like 15 forkhead-related protein FKHL15 thyroid transcription factor 2; forkhead box E2; Forkhead box protein E1; Forkhead box protein E2; forkhead, drosophila, homolog-like 15; Forkhead-related protein FKHL15; HFKH4; HFKL5; HNF-3/fork head-like protein 5; Thyroid transcription factor 2; TTF-2
Gene Aliases: FKHL15; FOXE1; FOXE2; HFKH4; HFKL5; NMTC4; TITF2; TTF-2; TTF2
UniProt ID: (Human) Q99526
Entrez Gene ID: (Human) 2304
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.