Invitrogen
Manufacturer Code:PA535145
Catalog # PIPA535145
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human FOXD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Forkhead box protein D1; Forkhead, drosophila, homolog-like 8; forkhead-like 8; forkhead-related activator 4; Forkhead-related protein FKHL8; Forkhead-related transcription factor 4; FOXD1 FKHL8 FREAC4 Forkhead box protein D1 Forkhead-related protein FKHL8 Forkhead-related transcription factor 4; FREAC-4
Gene Aliases: FKHL8; FOXD1; FREAC-4; FREAC4
UniProt ID: (Human) Q16676
Entrez Gene ID: (Human) 2297
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.