Proteintech
Manufacturer Code:230661AP150UL
Catalog # 50-173-3989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FOXC2 Fusion Protein Ag19378 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BF-3; Brain factor 3; FKHL14 Forkhead box protein C2 FOXC2 LD Mesenchyme fork head protein 1 MFH 1 MFH 1 protein MFH1 Transcription factor FKH 14; forkhead box C2 (MFH-1 mesenchyme forkhead 1); forkhead box C2 (MFH-1, mesenchyme forkhead 1); Forkhead box protein C2; forkhead homolog 14; forkhead, Drosophila, homolog-like 14; Forkhead-related protein FKHL14; HFH-BF-3; Mesenchyme fork head protein 1; mesenchyme forkhead 1; MFH-1 protein; MFH-1,mesenchyme forkhead 1; Transcription factor FKH-14
Gene Aliases: Fkh14; FKHL14; FOXC2; Hfhbf3; LD; MFH-1; MFH1
UniProt ID: (Human) Q99958, (Rat) Q63246, (Mouse) Q61850
Entrez Gene ID: (Human) 2303, (Rat) 171356, (Mouse) 14234
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.