Abnova
Manufacturer Code:H00002296M09
Catalog # 89-022-130
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E11 |
Immunogen | FOXC1 (NP_001444 464 a.a. ~ 553 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant FOXC1.
For Research Use Only
Protein Aliases: ARA; FKHL7; forkhead box C1 protein; Forkhead box protein C1; forkhead, drosophila, homolog-like 7; forkhead-related activator 3; Forkhead-related protein FKHL7; Forkhead-related transcription factor 3; forkhead/winged helix-like transcription factor 7; FREAC-3; FREAC3; IGDA; IHG1; IRID1; myeloid factor-delta
Gene Aliases: ARA; FKHL7; FOXC1; FREAC-3; FREAC3; IGDA; IHG1; IRID1; RIEG3
UniProt ID: (Human) Q12948
Entrez Gene ID: (Human) 2296
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.