Invitrogen
Manufacturer Code:PA518458
Catalog # PIPA518458
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (DAVKDKEEKDRLH) corresponding to the internal amino acids of FOXC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: forkhead box C1 protein; Forkhead box protein C1; forkhead, drosophila, homolog-like 7; forkhead-related activator 3; Forkhead-related protein FKHL7; Forkhead-related transcription factor 3; forkhead/winged helix-like transcription factor 7; FOXC1 forkhead box C1 HGNC:3800 ARA FKHL7 FREAC3 IGDA IHG1 IRID1 Forkhead drosophila homolog-like 7 forkhead (Drosophila)-like 7 forkhead-related activator 3 iridogoniodysgenesis type 1 myeloid factor-delta FREAC-3 RIEG3 forkhead/winged helix-like transcription factor 7; FREAC-3; myeloid factor-delta
Gene Aliases: ARA; FKHL7; FOXC1; FREAC-3; FREAC3; IGDA; IHG1; IRID1; RIEG3
UniProt ID: (Human) Q12948
Entrez Gene ID: (Human) 2296
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.