Invitrogen
Manufacturer Code:PA1807
Catalog # PIPA1807
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues A(423) V D D P L P D Y S L P(434) of human FOXC1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: congenital hydrocephalus; forkhead box C1 protein; Forkhead box protein C1; Forkhead box protein C1 FREAC-3; forkhead, drosophila, homolog-like 7; forkhead-related activator 3; Forkhead-related protein FKHL7; Forkhead-related transcription factor 3; forkhead/winged helix-like transcription factor 7; FREAC-3; Mesoderm/mesenchyme forkhead 1; MF-1; myeloid factor-delta; Transcription factor FKH-1
Gene Aliases: ARA; ch; fkh-1; Fkh1; FKHL7; FOXC1; FREAC-3; FREAC3; frkhda; IGDA; IHG1; IRID1; Mf1; Mf4; RIEG3
UniProt ID: (Human) Q12948, (Mouse) Q61572
Entrez Gene ID: (Human) 2296, (Mouse) 17300
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.