Invitrogen
Manufacturer Code:PA5101247
Catalog # PIPA5101247
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human FOXB1/2 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA159H20.4 C43 Fkh4 Fkh5 forkhead box B1 forkhead box B1a forkhead box B1b forkhead box B2 forkhead box protein B1 Forkhead box protein B2 forkhead homolog 4 Foxb1 Foxb1a Foxb1b Foxb2 Hfh-e5.1 HFKH-5 mesoderm/mesenchyme forkhead 3 Mf3 Transcription factor FKH-4 transcription factor FKH-5 Twh; forkhead box B1a; forkhead box B1b; Forkhead box protein B1; Forkhead box protein B2; forkhead homolog 4; mesoderm/mesenchyme forkhead 3; Transcription factor FKH-4; Transcription factor FKH-5
Gene Aliases: bA159H20.4; C43; Fkh4; FKH5; FOXB1; Foxb1a; Foxb1b; FOXB2; Hfh-e5.1; HFKH-5; Mf3; Twh
UniProt ID: (Human) Q99853, (Human) Q5VYV0, (Mouse) Q64732, (Mouse) Q64733
Entrez Gene ID: (Human) 27023, (Human) 442425, (Mouse) 64290, (Mouse) 14240
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.