Novus Biologicals
Manufacturer Code:NBP303931100UL
Catalog # NB095687
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human FOXB1 (NP_036314.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FKH5HFKH-5 forkhead box B1 forkhead box protein B1 Transcription factor FKH-5; Forkhead box protein B1; Transcription factor FKH-5
Gene Aliases: FKH5; FOXB1; HFKH-5
UniProt ID: (Human) Q99853
Entrez Gene ID: (Human) 27023
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.