Proteintech
Manufacturer Code:242851AP150UL
Catalog # 50-173-4277
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FOXB1 Fusion Protein Ag18079 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FKH5 forkhead box B1 Forkhead box protein B1 FOXB1 HFKH 5 Transcription factor FKH 5; forkhead box B1a; forkhead box B1b; Forkhead box protein B1; mesoderm/mesenchyme forkhead 3; Transcription factor FKH-5
Gene Aliases: C43; FKH5; FOXB1; Foxb1a; Foxb1b; Hfh-e5.1; HFKH-5; Mf3; Twh
UniProt ID: (Human) Q99853, (Mouse) Q64732
Entrez Gene ID: (Human) 27023, (Mouse) 64290
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.