Invitrogen
Manufacturer Code:PA1813
Catalog # PIPA1813
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues S(4) V K M E A H D L A E W S Y Y P E A G E(23) of mouse FOXA3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fork head-related protein FKH H3; Forkhead box A3; Forkhead box protein A3; hepatocyte nuclear factor 3 gamma (winged helix transcription factor); hepatocyte nuclear factor 3, gamma; Hepatocyte nuclear factor 3-gamma; HNF-3-gamma; HNF-3G; TCF-3G; Transcription factor 3G
Gene Aliases: FKHH3; FOXA3; Hnf-3g; HNF3G; Tcf-3g; TCF3G
UniProt ID: (Human) P55318, (Mouse) P35584, (Rat) P32183
Entrez Gene ID: (Human) 3171, (Mouse) 15377, (Rat) 25100
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.