Abnova
Manufacturer Code:H00003171K
Catalog # 89-205-925
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human FOXA3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human FOXA3 peptide using ARM Technology.
For Research Use Only
Protein Aliases: FKHH3 HNF3G MGC10179 TCF3G; Fork head-related protein FKH H3; Forkhead box protein A3; hepatocyte nuclear factor 3, gamma; Hepatocyte nuclear factor 3-gamma; HNF-3-gamma; HNF-3G; TCF-3G; Transcription factor 3G
Gene Aliases: FKHH3; FOXA3; HNF3G; TCF3G
UniProt ID: (Human) P55318
Entrez Gene ID: (Human) 3171
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.