Abnova
Manufacturer Code:MAB20056
Catalog # 89-336-456
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | AAAH-6 |
Immunogen | A synthetic peptide corresponding to human FOXA2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C for one year. After reconstitution at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. |
Rabbit Monoclonal Antibody.
Protein Aliases: Forkhead box protein A2; hepatic nuclear factor-3-beta; hepatocyte nuclear factor 3, beta; Hepatocyte nuclear factor 3-beta; HNF-3-beta; HNF-3B; HNF3B MGC19807 TCF3B; TCF-3B; Transcription factor 3B
Gene Aliases: FOXA2; HNF3B; TCF3B
UniProt ID: (Human) Q9Y261
Entrez Gene ID: (Human) 3170
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.