Invitrogen
Manufacturer Code:PA579281
Catalog # PIPA579281
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human FOXA1 recombit protein (Position: M1-T55). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: fork head domain; Forkhead box protein A1; hepatocyte nuclear factor 3 alpha (winged helix transcription factor); hepatocyte nuclear factor 3, alpha; Hepatocyte nuclear factor 3-alpha; Hepatocyte nuclear factor 3-alpha HNF-3-alpha HNF-3A Forkhead box protein A1 Transcription factor 3A TCF-3A FOXA1 HNF3A TCF3A; HNF-3-alpha; HNF-3A; TCF-3A; Transcription factor 3A
Gene Aliases: FOXA1; Hnf-3a; HNF3A; Tcf-3a; TCF3A
UniProt ID: (Human) P55317, (Mouse) P35582, (Rat) P23512
Entrez Gene ID: (Human) 3169, (Mouse) 15375, (Rat) 25098
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.