OriGene
Manufacturer Code:CF804303
Catalog # 50-166-6927
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2G5 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-255 of human FOSB produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: activator protein 1; AP-1 G0S3 GOS3 GOSB; FBJ murine osteosarcoma viral oncogene homolog B; FosB proto-oncogene, AP-1 trancription factor subunit; G0/G1 switch regulatory protein 3; Protein fosB
Gene Aliases: AP-1; FOSB; G0S3; GOS3; GOSB
UniProt ID: (Human) P53539
Entrez Gene ID: (Human) 2354
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.