Proteintech
Manufacturer Code:665901IG150UL
Catalog # 50-173-7008
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1G2C5 |
Immunogen | FOS Fusion Protein Ag24340 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: activator protein 1; AP 1 C FOS Cellular oncogene fos FOS G0S7 Proto oncogene c Fos; cellular oncogene c-fos; Cellular oncogene fos; FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS); FBJ murine osteosarcoma viral oncogene homolog; Fos proto-oncogene, AP-1 trancription factor subunit; G0/G1 switch regulatory protein 7; Proto-oncogene c-Fos; proto-oncogene protein c-fos
Gene Aliases: AP-1; C-FOS; cFos; D12Rfj1; FOS; G0S7; p55
UniProt ID: (Human) P01100, (Mouse) P01101
Entrez Gene ID: (Human) 2353, (Mouse) 14281
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.