Invitrogen
Manufacturer Code:MA524803
Catalog # PIMA524803
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 12A5 |
Immunogen | CD3 mAb-stimulated CD4/CD25 T cells. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Folate receptor 4; folate receptor 4 (delta); folate receptor 4 (delta) homolog (mouse) Folbp3 probable folate receptor delta; Folate receptor delta; Folate-binding protein 3; FR-delta; IZUMO1 receptor protein JUNO; probable folate receptor delta; Sperm-egg fusion protein Juno
Gene Aliases: 0910001L11Rik; C86255; Folbp3; Folr4; Izumo1r; Juno
UniProt ID: (Mouse) Q9EQF4
Entrez Gene ID: (Mouse) 64931
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.